4,5-Dichloro-2,3-dihydro-1H-inden-1-one ≥97%

Supplier: Ambeed
Warning

A284452-5G A284452-1G
77612-634EA 277.07 USD
77612-634 77683-854
4,5-Dichloro-2,3-dihydro-1H-inden-1-one ≥97%
4,5-Dichloro-2,3-dihydro-1H-inden-1-one

Precaución: For research use only.

Formula: C₉H₆Cl₂O
MW: 201.04 g/mol
MDL Number: MFCD13190286
CAS Number: 69392-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR