UNC0631 ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(1-(Cyclohexylmethyl)piperidin-4-yl)-2-(4-isopropyl-1,4-diazepan-1-yl)-6-methoxy-7-(3-(piperidin-1-yl)propoxy)quinazolin-4-amine, UNC 0631

A284379-1MG A284379-10MG A284379-5MG
77616-214EA 33.99 USD
77616-214 77746-521 77746-522
UNC0631 ≥98%
UNC0631

Precaución: for Research Use Only

Formula: C₃₇H₆₁N₇O₂
MW: 635.926 g/mol
MDL Number: MFCD22683862
CAS Number: 1320288-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR