7-Chloro-2,3-dihydroquinolin-4(1H)-one ≥95%

Supplier: Ambeed
Warning

A284293-250MG A284293-100MG
77685-807EA 106 USD
77685-807 77892-566
7-Chloro-2,3-dihydroquinolin-4(1H)-one ≥95%
7-Chloro-2,3-dihydroquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MDL Number: MFCD00795494
CAS Number: 21617-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR