tert-Butyl (4-bromobutan-2-yl)carbamate ≥95%

Supplier: Ambeed
Warning

A284262-1G A284262-250MG
77901-304EA 126 USD
77901-304 77900-542
tert-Butyl (4-bromobutan-2-yl)carbamate ≥95%
tert-Butyl (4-bromobutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈BrNO₂
MDL Number: MFCD22576327
CAS Number: 128412-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR