Methyl-3-(4-chlorosulfonyl)phenylpropionate ≥98%

Supplier: Ambeed
Danger

Synonyms: Methyl 3-(4-(chlorosulphonyl)phenyl)propanoate

A284253-1G A284253-5G
77668-258EA 28 USD
77668-258 77668-260
Methyl-3-(4-chlorosulfonyl)phenylpropionate ≥98%
Methyl-3-(4-chlorosulfonyl)phenylpropionate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₄S
MW: 262.7099 g/mol
MDL Number: MFCD02093942
CAS Number: 374537-95-8
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR