Sodium cholate ≥98%

Supplier: Ambeed
Warning

A284196-100G A284196-500G A284196-25G
77626-452EA 82.4 USD
77626-452 77626-454 77879-549
Sodium cholate ≥98%
Sodium cholate

Precaución: for Research Use Only

Formula: C₂₄H₃₉NaO₅
MDL Number: MFCD00150749
CAS Number: 361-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR