tert-Butyl (4-amino-3-methylphenyl)carbamate ≥97%

Supplier: Ambeed
Warning

A284045-100MG A284045-1G A284045-250MG
77892-563EA 30 USD
77892-563 77892-564 77892-565
tert-Butyl (4-amino-3-methylphenyl)carbamate ≥97%
tert-Butyl (4-amino-3-methylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD05663969
CAS Number: 325953-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR