3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile, 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

A283891-100MG A283891-1G
77616-210EA 47.38 USD
77616-210 77616-212
3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester ≥98%
3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₃BBrNO₂
MW: 293.952 g/mol
MDL Number: MFCD08669549
CAS Number: 883899-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR