Potassium hexabromoplatinate ≥98%

Supplier: Ambeed
Danger

A283854-250MG A283854-100MG A283854-1G
77668-254EA 49 USD
77668-254 77879-546 77879-547
Potassium hexabromoplatinate ≥98%
Potassium hexabromoplatinate

Precaución: for Research Use Only

Formula: Br₆K₂Pt
MDL Number: MFCD00011377
CAS Number: 16920-93-7
UN: 3290
ADR: 6.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR