6-Methoxy-2-azaspiro[3.3]heptane hydrochloride ≥98%

Supplier: Ambeed
Warning

A283338-100MG A283338-1G A283338-250MG
77616-204EA 47.38 USD
77616-204 77616-206 77616-208
6-Methoxy-2-azaspiro[3.3]heptane hydrochloride ≥98%
6-Methoxy-2-azaspiro[3.3]heptane hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO·ClH
MW: 163.645 g/mol
MDL Number: MFCD28501414
CAS Number: 1638761-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR