2-((1,3-Dioxoisoindolin-2-yl)oxy)acetic acid ≥97%

Supplier: Ambeed
Warning

A282779-100MG A282779-5G
77746-515EA 143 USD
77746-515 77746-516
2-((1,3-Dioxoisoindolin-2-yl)oxy)acetic acid ≥97%
2-((1,3-Dioxoisoindolin-2-yl)oxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₅
MDL Number: MFCD11850832
CAS Number: 134724-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR