tert-Butyl ((3-aminobicyclo[1.1.1]pentan-1-yl)methyl)carbamate ≥98%

Supplier: Ambeed
Danger

Synonyms: tert-butyl N-({3-aminobicyclo[1.1.1]pentan-1-yl}methyl)carbamate

A282410-1G A282410-50MG
77685-798EA 1792 USD
77685-798 77685-799
tert-Butyl ((3-aminobicyclo[1.1.1]pentan-1-yl)methyl)carbamate ≥98%
tert-Butyl ((3-aminobicyclo[1.1.1]pentan-1-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.289 g/mol
MDL Number: MFCD27987306
CAS Number: 1630906-49-8
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR