7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2-(3,4-dihydroxyphenyl)-5-hydroxy-4H-chromen-4-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Lonicerin

A282319-10MG A282319-50MG
77746-510EA 195 USD
77746-510 77746-511
7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2-(3,4-dihydroxyphenyl)-5-hydroxy-4H-chromen-4-one ≥98%
7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2-(3,4-dihydroxyphenyl)-5-hydroxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₇H₃₀O₁₅
MW: 594.52 g/mol
CAS Number: 25694-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR