3-Bromo-2-fluorobenzenesulfonyl chloride ≥98%, Ambeed.Inc

Supplier: Ambeed
Danger

A282052-5G A282052-1G
77683-849EA 153 USD
77683-849 77892-558
3-Bromo-2-fluorobenzenesulfonyl chloride ≥98%, Ambeed.Inc
3-Bromo-2-fluorobenzenesulfonyl chloride

Precaución: For research use only.

Formula: C₆H₃BrClFO₂S
MDL Number: MFCD13185362
CAS Number: 1214372-19-6
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR