Chloro(1,5-cyclooctadiene)methylpalladium(II) ≥98%

Supplier: Ambeed
Danger

A281583-1G A281583-250MG
77746-507EA 333 USD
77746-507 77746-508
Chloro(1,5-cyclooctadiene)methylpalladium(II) ≥98%
Chloro(1,5-cyclooctadiene)methylpalladium(II)

Precaución: for Research Use Only

Formula: C₉H₁₀ClPd
MDL Number: MFCD16621448
CAS Number: 63936-85-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR