LCQ-908 ≥98%

Supplier: Ambeed
Warning

Synonyms: Pradigastat, 2-(trans-4-(4-(5-((6-(Trifluoromethyl)pyridin-3-yl)amino)pyridin-2-yl)phenyl)cyclohexyl)acetic acid

A281007-10MG A281007-1MG A281007-5MG
77616-190EA 145.23 USD
77616-190 77616-192 77616-194
LCQ-908 ≥98%
LCQ-908

Precaución: for Research Use Only

Formula: C₂₅H₂₄F₃N₃O₂
MW: 455.47 g/mol
MDL Number: MFCD25562905
CAS Number: 956136-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR