(5-(Thiophen-2-yl)isoxazol-3-yl)methanol ≥97%

Supplier: Ambeed
Warning

Synonyms: [5-(2-Thienyl)-3-isoxazolyl]methanol

A280566-100MG A280566-250MG
77601-236EA 42.03 USD
77601-236 77626-444
(5-(Thiophen-2-yl)isoxazol-3-yl)methanol ≥97%
(5-(Thiophen-2-yl)isoxazol-3-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₇NO₂S
MW: 181.21 g/mol
MDL Number: MFCD03086162
CAS Number: 194491-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR