2,2'-Dibromoacetophenone ≥95%

Supplier: Ambeed
Danger

Synonyms: 2-Bromophenacyl bromide, 2-Bromo-1-(2-bromophenyl)ethanone

A279884-25G A279884-5G
77746-502EA 352 USD
77746-502 77746-503
2,2'-Dibromoacetophenone ≥95%
2,2'-Dibromoacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Br₂O
MW: 277.9406 g/mol
MDL Number: MFCD00044660
CAS Number: 49851-55-0
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR