5-Fluorobenzo[b]furan ≥95%

Supplier: Ambeed
Warning

A279766-100MG A279766-250MG A279766-1G A279766-5G
77626-436EA 38.11 USD
77626-436 77626-438 77877-928 77877-929
5-Fluorobenzo[b]furan ≥95%
5-Fluorobenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₅FO
MDL Number: MFCD08236757
CAS Number: 24410-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR