LXS-196 ≥99%

Supplier: Ambeed
Warning

Synonyms: NVP-LXS196, 3-amino-N-(3-(4-amino-4-methylpiperidin-1-yl)pyridin-2-yl)-6-(3-(trifluoromethyl)pyridin-2-yl)pyrazine-2-carboxamide, Darovasertib

A279706-1MG A279706-10MG A279706-5MG
77616-186EA 32.96 USD
77616-186 77748-542 77748-543
LXS-196 ≥99%
LXS-196

Precaución: for Research Use Only

Formula: C₂₂H₂₃F₃N₈O
MW: 472.5 g/mol
MDL Number: MFCD31544328
CAS Number: 1874276-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR