4-Hydroxy-3,4-dihydronaphthalen-1(2H)-one ≥95%

Supplier: Ambeed
Warning

A279533-5G A279533-1G A279533-250MG
77571-738EA 692 USD
77571-738 77872-341 77872-342
4-Hydroxy-3,4-dihydronaphthalen-1(2H)-one ≥95%
4-Hydroxy-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD11036279
CAS Number: 21032-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR