3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid ≥98%

Supplier: Ambeed
Warning

A279490-100G A279490-5G
77672-436EA 359 USD
77672-436 77881-760
3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid ≥98%
3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆O₆S
MDL Number: MFCD00966477
CAS Number: 18361-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR