AZD8835 ≥98%

Supplier: Ambeed
Warning

A279294-10MG A279294-5MG
77685-785EA 96 USD
77685-785 77685-786
AZD8835 ≥98%
AZD8835

Precaución: for Research Use Only

Formula: C₂₂H₃₁N₉O₃
MDL Number: MFCD28902194
CAS Number: 1620576-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR