Fmoc-isoGln-OH ≥97%

Supplier: Ambeed
Warning

Synonyms: (S)-4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-5-amino-5-oxopentanoic acid, Fmoc-Glu-NH2, Fmoc-Glu-NH₂

A279083-1G A279083-250MG
77885-745EA 16 USD
77885-745 77885-746
Fmoc-isoGln-OH ≥97%
Fmoc-isoGln-OH

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₅
MDL Number: MFCD01861342
CAS Number: 288149-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR