3,4-Dibromobenzonitrile ≥95%

Supplier: Ambeed
Warning

A279000-5G A279000-1G
77612-612EA 62.83 USD
77612-612 77901-307
3,4-Dibromobenzonitrile ≥95%
3,4-Dibromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Br₂N
MDL Number: MFCD09800818
CAS Number: 188984-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR