1-(5-(Furan-2-yl)-1,3,4-oxadiazol-2-yl)-3-(pyridin-2-ylmethyl)urea ≥98%

Supplier: Ambeed
Warning

Synonyms: NK-252

A278611-10MG A278611-1MG A278611-5MG
77616-174EA 45.32 USD
77616-174 77616-176 77616-178
1-(5-(Furan-2-yl)-1,3,4-oxadiazol-2-yl)-3-(pyridin-2-ylmethyl)urea ≥98%
1-(5-(Furan-2-yl)-1,3,4-oxadiazol-2-yl)-3-(pyridin-2-ylmethyl)urea

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₅O₃
MW: 285.25 g/mol
MDL Number: MFCD28009512
CAS Number: 1414963-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR