Methyl 5-bromo-3-hydroxy-2-methylbenzoate ≥97%

Supplier: Ambeed
Warning

A278294-10G A278294-5G A278294-1G A278294-25G
77571-726EA 899.19 USD
77571-726 77571-728 77598-394 77748-535
Methyl 5-bromo-3-hydroxy-2-methylbenzoate ≥97%
Methyl 5-bromo-3-hydroxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD27987185
CAS Number: 1492040-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR