1-Boc-5-methoxyindole-2-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-5-methoxy-2-indolylboronic acid, (1-(tert-Butoxycarbonyl)-5-methoxy-1H-indol-2-yl)boronic acid

A277843-1G A277843-5G
77571-718EA 36 USD
77571-718 77872-337
1-Boc-5-methoxyindole-2-boronic acid ≥98%
1-Boc-5-methoxyindole-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₅
MW: 291.11
MDL Number: MFCD04039006
CAS Number: 290331-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR