6,6'-Bis(bromomethyl)-2,2'-bipyridine ≥98%

Supplier: Ambeed
Danger

A277669-1G A277669-100MG A277669-250MG
77612-610EA 193.64 USD
77612-610 77683-839 77683-840
6,6'-Bis(bromomethyl)-2,2'-bipyridine ≥98%
6,6'-Bis(bromomethyl)-2,2'-bipyridine

Precaución: For research use only.

Formula: C₁₂H₁₀Br₂N₂
MW: 342.03 g/mol
MDL Number: MFCD27920801
CAS Number: 96517-97-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR