Bis-PEG1-acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 3,3'-Oxydipropanoic acid

A277487-1G A277487-100MG A277487-250MG
77571-716EA 126.69 USD
77571-716 77612-606 77685-781
Bis-PEG1-acid ≥95%
Bis-PEG1-acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₅
MW: 162.141 g/mol
MDL Number: MFCD06208032
CAS Number: 5961-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR