((2R,3R,4S,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-3,5-dihydroxy-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)methyl (E)-3-(3,4-dihydroxyphenyl)acrylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Isoacteoside

A276968-100MG A276968-250MG
77892-542EA 317 USD
77892-542 77892-543
((2R,3R,4S,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-3,5-dihydroxy-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)methyl (E)-3-(3,4-dihydroxyphenyl)acrylate ≥98%
((2R,3R,4S,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-3,5-dihydroxy-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)methyl (E)-3-(3,4-dihydroxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₂₉H₃₆O₁₅
MDL Number: MFCD06798947
CAS Number: 61303-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR