4-(Dimethoxymethyl)-2-methylpyrimidine ≥97%

Supplier: Ambeed
Warning

A276654-5G A276654-1G
77616-166EA 95.79 USD
77616-166 77748-530
4-(Dimethoxymethyl)-2-methylpyrimidine ≥97%
4-(Dimethoxymethyl)-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.19 g/mol
MDL Number: MFCD00085137
CAS Number: 175277-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR