Nampt-IN-1 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-2-methyl-N-[1,2,3,4-tetrahydro-2-[2-(3-pyridinyloxy)acetyl]-6-isoquinolinyl]-1-propanesulphonamide, 2-Hydroxy-2-methyl-N-(2-(2-(pyridin-3-yloxy)acetyl)-1,2,3,4-tetrahydroisoquinolin-6-yl)propane-1-sulphonamide

A276413-10MG A276413-1MG A276413-5MG
77616-160EA 119.48 USD
77616-160 77616-162 77616-164
Nampt-IN-1 ≥98%
Nampt-IN-1

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₃O₅S
MW: 419.49 g/mol
MDL Number: MFCD28963946
CAS Number: 1698878-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR