Di-tert-butyl hydrogen phosphate ≥95%

Supplier: Ambeed
Danger

A275719-5G A275719-1G
77571-702EA 211.15 USD
77571-702 77601-220
Di-tert-butyl hydrogen phosphate ≥95%
Di-tert-butyl hydrogen phosphate

Precaución: for Research Use Only

Formula: C₈H₁₉O₄P
MW: 210.21 g/mol
MDL Number: MFCD08443810
CAS Number: 33494-81-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR