4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one ≥95%

Supplier: Ambeed
Warning

A275638-1G A275638-25G
77877-917EA 23 USD
77877-917 77877-918
4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one ≥95%
4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MDL Number: MFCD00085024
CAS Number: 67751-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR