Methyl cis-3-hydroxymethylcyclopentane-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A275233-100MG A275233-250MG
77901-942EA 65 USD
77901-942 77901-135
Methyl cis-3-hydroxymethylcyclopentane-1-carboxylate ≥98%
Methyl cis-3-hydroxymethylcyclopentane-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD06799098
CAS Number: 58240-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR