Ethyl 4-chloro-1H-indole-2-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-Chloro-2-indolecarboxylate

A274924-5G A274924-250MG
77593-698EA 147.6 USD
77593-698 77668-202
Ethyl 4-chloro-1H-indole-2-carboxylate ≥98%
Ethyl 4-chloro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO₂
MW: 223.656 g/mol
MDL Number: MFCD09264531
CAS Number: 53590-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR