3-Bromo-2-methoxyquinoline ≥95%

Supplier: Ambeed
Warning

A273699-100MG A273699-1G A273699-250MG
77890-494EA 39 USD
77890-494 77890-495 77890-496
3-Bromo-2-methoxyquinoline ≥95%
3-Bromo-2-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD12827492
CAS Number: 222317-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR