3-Chloro-4-morpholinophenylboronic Acid ≥98%

Supplier: Ambeed
Warning

A273696-100MG A273696-1G A273696-250MG
77872-324EA 76 USD
77872-324 77872-325 77872-326
3-Chloro-4-morpholinophenylboronic Acid ≥98%
3-Chloro-4-morpholinophenylboronic Acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₃
MDL Number: MFCD16660219
CAS Number: 1426246-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR