H-D-Lys(Boc)-OH ≥95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-6-((tert-butoxycarbonyl)amino)hexanoic acid, Nepsilon-Boc-D-lysine

A273424-1G A273424-250MG
77685-773EA 25 USD
77685-773 77685-774
H-D-Lys(Boc)-OH ≥95%
H-D-Lys(Boc)-OH

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₄
MW: 246.307 g/mol
MDL Number: MFCD00076958
CAS Number: 31202-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR