Tropone ≥96%

Supplier: Ambeed
Warning

Synonyms: Cyclohepta-2,4,6-trien-1-one, 2,4,6-Cycloheptatrienone, 2,4,6-Cycloheptatrien-1-one

A272890-1G A272890-250MG A272890-5G
77601-182EA 46.13 USD
77601-182 77601-184 77668-190
Tropone ≥96%
Tropone

Precaución: for Research Use Only

Formula: C₇H₆O
MW: 106.12 g/mol
MDL Number: MFCD00014331
CAS Number: 539-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR