1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: 7-Amino-N-methyl-1,2,3,4-tetrahydroquinoline hydrochloride

A272846-1G A272846-250MG A272846-10G
77593-680EA 111.73 USD
77593-680 77593-682 77668-188
1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride ≥95%
1-Methyl-1,2,3,4-tetrahydroquinolin-7-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅ClN₂
MW: 198.69 g/mol
MDL Number: MFCD09839668
CAS Number: 927684-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR