4-Methoxy-6-methyl-1H-indole ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A272704-100MG A272704-1G A272704-250MG
77683-830EA 66 USD
77683-830 77885-723 77885-724
4-Methoxy-6-methyl-1H-indole ≥95%, Ambeed.Inc
4-Methoxy-6-methyl-1H-indole

Precaución: For research use only.

Formula: C₁₀H₁₁NO
MDL Number: MFCD07781526
CAS Number: 885522-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR