2-Chlorobenzo[d]thiazol-6-amine ≥97%

Supplier: Ambeed
Warning

A272625-1G A272625-250MG
77571-680EA 30 USD
77571-680 77616-150
2-Chlorobenzo[d]thiazol-6-amine ≥97%
2-Chlorobenzo[d]thiazol-6-amine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂S
MDL Number: MFCD08276901
CAS Number: 2406-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR