7-Methyl-1,3-dihydro-2H-indol-2-one ≥98%

Supplier: Ambeed
Warning

A272368-100MG A272368-1G A272368-250MG
77877-908EA 45 USD
77877-908 77877-909 77877-910
7-Methyl-1,3-dihydro-2H-indol-2-one ≥98%
7-Methyl-1,3-dihydro-2H-indol-2-one

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD02179609
CAS Number: 3680-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR