3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R,2R)-2-(dimethylamino)-1,2-diphenylethyl)amino)cyclobut-3-ene-1,2-dione ≥98%

Supplier: Ambeed
Warning

A272342-100MG A272342-250MG
77746-479EA 45 USD
77746-479 77746-480
3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R,2R)-2-(dimethylamino)-1,2-diphenylethyl)amino)cyclobut-3-ene-1,2-dione ≥98%
3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R,2R)-2-(dimethylamino)-1,2-diphenylethyl)amino)cyclobut-3-ene-1,2-dione

Precaución: for Research Use Only

Formula: C₂₈H₂₃F₆N₃O₂
MW: 547.49 g/mol
CAS Number: 1223105-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR