2-(5-Methyl-1H-pyrazol-3-yl)pyridine ≥95%

Supplier: Ambeed
Warning

A272313-250MG A272313-1G
77881-740EA 39 USD
77881-740 77881-739
2-(5-Methyl-1H-pyrazol-3-yl)pyridine ≥95%
2-(5-Methyl-1H-pyrazol-3-yl)pyridine

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD11840239
CAS Number: 19959-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR