2-Methyl-6-(phenylethynyl)pyridine ≥98%

Supplier: Ambeed
Warning

A271724-250MG A271724-100MG A271724-1G
77601-162EA 23.58 USD
77601-162 77668-184 77668-186
2-Methyl-6-(phenylethynyl)pyridine ≥98%
2-Methyl-6-(phenylethynyl)pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MDL Number: MFCD03787983
CAS Number: 96206-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR