2-Amino-4-fluorobenzeneboronic acid pinacol ester ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzenamine, 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 2-Amino-4-fluorophenylboronic acid pinacol ester, 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
A271540-1G
A271540-100MG
77571-674EA
289.43
USD
77571-674
77683-827
2-Amino-4-fluorobenzeneboronic acid pinacol ester ≥95%
2-Amino-4-fluorobenzeneboronic acid pinacol ester
Precaución: For research use only.
Formula:
C₁₂H₁₇BFNO₂ MW: 237.08 g/mol |
MDL Number:
MFCD12025504 CAS Number: 1309982-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...