1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (1-(Tetrahydro-2H-pyran-2-yl)-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid

A271399-100MG A271399-1G A271399-250MG
77885-718EA 37 USD
77885-718 77885-719 77885-720
1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid ≥98%
1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BF₃N₂O₃
MW: 264.01 g/mol
MDL Number: MFCD17214248
CAS Number: 1141878-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR